Learn More
Bellco Glass, Inc. Snap-Ring Clear Vial,2.0mL Big Mouth-11mm Crimp Top
SDP
Catalog No. 501945360
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Developed for use with Hewlett Packard and other auto-samplers. Vials accept 11mm snap-ring closures. Patented snap-seal design for positive seal. MADE IN THE USA. Size: 12x32mm Snap-ring size: 11mm
Catalog No. 50-194-5360 Supplier Bellco Glass, Inc. Supplier No. 207000070
Manufactured from type 1 class a Borosilicate glass. Developed for use with Hewlett Packard and other auto-samplers. Vials accept 11mm snap-ring closures. Patented snap-seal design for positive seal. MADE IN THE USA. Size: 12x32mm Snap-ring size: 11mm