Learn More
Waters Corp Snap-Ring Pliers, C Ring Removal, Plunger Installation
Catalog No. 502392836 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Snap-Ring Pliers are designed to facilitate the removal and installation of the C ring, which secures the sapphire plunger within pump heads.
  • Helps to remove the C ring that holds the sapphire plunger in place
  • Assists in the installation of the C ring that holds the sapphire plunger in place
  • Suitable for use with pump heads
Catalog No. 50-239-2836 Supplier Waters Corp Supplier No. WAT025263
The Snap-Ring Pliers are designed to facilitate the removal and installation of the C ring, which secures the sapphire plunger within pump heads.
  • Helps to remove the C ring that holds the sapphire plunger in place
  • Assists in the installation of the C ring that holds the sapphire plunger in place
  • Suitable for use with pump heads