Learn More
Andwin Scientific SNAP STRP NAT.PCR TBS 120 OF 8
SDP
Catalog No. NC9860800
Encompass_Preferred
The 0.2 mL GeneMate SnapStrip 8-Strip Standard PCR Tubes feature individually attached dome caps and ultra-thin wall that permits efficient heat transfer. This product conveniently allows users to construct a strip of less than 8 tubes with caps by simply breaking the strip where desired. It is sold in a natural color in a quantity of 120.
Catalog No. NC9860800 Supplier Andwin Scientific Supplier No. T30351
The 0.2 mL GeneMate SnapStrip 8-Strip Standard PCR Tubes feature individually attached dome caps and ultra-thin wall that permits efficient heat transfer. This product conveniently allows users to construct a strip of less than 8 tubes with caps by simply breaking the strip where desired. It is sold in a natural color in a quantity of 120.