Learn More
New England Biolabs, Inc. SNAP-Surface 488 – 50 nmol
SDP
Catalog No. 508112055
Encompass
SNAP-Surface 488 is a photostable fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living cells.
Catalog No. 50-811-2055 Supplier New England Biolabs, Inc. Supplier No. S9124S
SNAP-Surface 488 is a photostable fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living cells.