Learn More
New England Biolabs, Inc. SNAP-Surface 549 – 50 nmol
SDP
Catalog No. 508112054
Encompass
SNAP-Surface 549 is a photostable red fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living cells.
Catalog No. 50-811-2054 Supplier New England Biolabs, Inc. Supplier No. S9112S
SNAP-Surface 549 is a photostable red fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living cells.