Learn More
New England Biolabs, Inc. SNAP-Surface 594 – 50 nmol
SDP
Catalog No. 508112060
Encompass
SNAP-Surface 594 is a photostable fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living or fixed cells.
Catalog No. 50-811-2060 Supplier New England Biolabs, Inc. Supplier No. S9134S
SNAP-Surface 594 is a photostable fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living or fixed cells.