Learn More
New England Biolabs, Inc. SNAP-Surface Alexa Fluor 488 – 50 nmol
SDP
Catalog No. 508112057
Encompass
SNAP-Surface Alexa Fluor 488 is a photostable green fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living cells.
Catalog No. 50-811-2057 Supplier New England Biolabs, Inc. Supplier No. S9129S
SNAP-Surface Alexa Fluor 488 is a photostable green fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living cells.