Learn More
New England Biolabs, Inc. SNAP-Surface Alexa Fluor 546 – 50 nmol
SDP
Catalog No. 508112059
Encompass
SNAP-Surface Alexa Fluor 546 is a photostable fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living cells.
Catalog No. 50-811-2059 Supplier New England Biolabs, Inc. Supplier No. S9132S
SNAP-Surface Alexa Fluor 546 is a photostable fluorescent substrate that can be used to label SNAP-tag fusion proteins in solution or on the surface of living cells.