Learn More
New England Biolabs, Inc. SNAP-Surface Block – 200 nmol
SDP
Catalog No. 508112068
Encompass
SNAP-Surface Block (C8 propanoic acid benzylguanine, CBG) is a nonfluorescent compound that blocks the reactivity of the SNAP-tag in solution or on the surface of living cells.
Catalog No. 50-811-2068 Supplier New England Biolabs, Inc. Supplier No. S9143S
SNAP-Surface Block (C8 propanoic acid benzylguanine, CBG) is a nonfluorescent compound that blocks the reactivity of the SNAP-tag in solution or on the surface of living cells.