Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Snap Tm Reagent

Catalog No. 7988861G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-888-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 79-888-61G Supplier Sigma Aldrich Supplier No. 7988861G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

SnAP (Sn amino protocol) reagents have been developed by the Bode group. SnAP reagents are air and moisture stable. They can be prepared from simple reactions with cheap raw materials. SnAP 2-Spiro-(4-Pip) M, also referred to as SnAP 2-spiro-(4-piperdine) morpholine, is a regioisomeric SnAP reagent. This reagent can be prepared from N-Boc piperidone.SnAP TM Reagent may be used to synthesize 3-substituted thiomorpholines from aldehydes. It may also be used in the preparation of unprotected, saturated N-heterocyclic compounds.Technology spotlight: SnAP Reagents Professor product portal: Jeffrey Bode Research GroupSnAP Reagents for the Synthesis of Piperazines and MorpholinesSnAP reagents for the one-step synthesis of medium-ring saturated N-heterocycles from aldehydesSnAP Reagents for a Cross-Coupling Approach to the One-Step Synthesis of Saturated N-HeterocyclesBespoke SnAP Reagents for the Synthesis of C-Substituted Spirocyclic and Bicyclic Saturated N-Heterocycles

Specifications

CAS 1577233-73-8
Molecular Formula C21H46N2O2Sn
Refractive Index n/D 1.512
Linear Formula C21H46N2O2Sn
Quantity 1 g
Synonym tert-Butyl (3-aminopropyl)((tributylstannyl)methyl)carbamate
Molecular Weight (g/mol) 477.31 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.