Learn More
StatLab Snap Top Empty Specimen Container
SDP
Catalog No. 21033348
Change view
Click to view available options
Capacity (English):
15 oz.
32 oz.
64 oz.
80 oz.
128 oz.
165 oz.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
21-033-348 15 oz.
21-033-349 32 oz.
21-033-350 64 oz.
21-033-351 80 oz.
21-033-352 165 oz.
21-033-353 128 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 21-033-348 Supplier StatLab Supplier No. CT3702
Add to Cart
Edge
Add to Cart
Designed for safe leak-resistant transport and storage of fluids
  • StatLab containers are designed for safe, leak-resistant transport and storage of fluids
  • They are graduated and made of durable clarified polypropylene
  • Larger snap-top containers are made of HDPE
  • Lids included
Specifications
Product Type Specimen Container
Closure Included Yes
Graduated Yes
Material HDPE
Shape Cylindrical
Sterility Sterile
Capacity (English) 15 oz.