Learn More
StatLab Snap Top Empty Specimen Container
Description
- StatLab containers are designed for safe, leak-resistant transport and storage of fluids
- They are graduated and made of durable clarified polypropylene
- Larger snap-top containers are made of HDPE
- Lids included
Specifications
Specifications
| Product Type | Specimen Container |
| Closure Included | Yes |
| Graduated | Yes |
| Material | HDPE |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Capacity (English) | 80 oz. |