Learn More
Azer Scientific Snap Top Pathology Container
Description
- Our snap top pathology containers are sturdy, light-weight, and shatter resistant Manufactured from translucent, high-density polyethylene
- Snap-on lid creates a secure seal preventing leakage
- Pathology containers are resistant to acids, fixatives and alcohols
- Non-sterile
- Excellent resistance to acids, fixatives, and alcohols
- Sturdy, light-weight, and shatter resistant
- Snap-on lid
Specifications
Specifications
| Product Type | Specimen Container |
| Closure Included | Yes |
| Graduated | Yes |
| Material | HDPE |
| Shape | Rectangular |
| Sterility | Non-sterile |
| Quantity | 85 oz. |