Learn More
Cardinal Health™ Snap-up Frock with Tyvek™
Catalog No. 191302124 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
2X-Large
4X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-2124 4X-Large
19-130-2013 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.
Catalog No. 19-130-2124 Supplier Cardinal Health™ Supplier No. 9347
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.
Full-length frock features snaps to the neck for maximum protection.
  • Features adjustable high tab collar
  • No pockets to trap dirt or lint
  • Basic style offers standard fit with set-in sleeves
  • Premier style offers raglan sleeves with a more generous cut
  • Packed in double poly liners to ensure cleanliness
TRUSTED_SUSTAINABILITY
Specifications
Size 4X-Large
Material Tyvek
Color White