Learn More
Aviva Systems Biology Corp SNAP25 Antibody - C-terminal region (AVARP13090_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13090_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNAP25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse SNAP25. Alternate names: sp, Bdr, SNAP-25ApplicationWB. Species reactivity: Dog, Guinea Pig, Rat
Catalog No. 50-014-62637