Learn More
Aviva Systems Biology Corp SNAPC1 Antibody - C-terminal region (ARP38227_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38227_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SNAPC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SNAPC1. Alternate names: PTFgamma, SNAP43ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56572