Learn More
Aviva Systems Biology Corp SNAPC2 Antibody - N-terminal region (ARP31318_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31318_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNAPC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SNAPC2. Alternate names: PTFdelta, SNAP45, PTFDELTAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54194