Learn More
Aviva Systems Biology Corp SNAPC3 Antibody - N-terminal region (ARP34174_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34174_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SNAPC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SNAPC3. Alternate names: SNAP50, PTFbetaApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55281