Learn More
Applied Biosystems™ SNaPshot™ Multiplex Kit
Description
Order Info
Each
Specifications
Specifications
| Accession Number | Capillary, 61cm Capillary Length, Sequencing, Room Temperature, For 310 Genetic Analyzer |
| Description | SNaPshot™ Multiplex Kit |
| No. of Reactions | 5000 |
For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.