Learn More
Research Products International Corp SnapStrip II Tubes with Caps, Low Profile, Natural, 120 Strips, 960 Tubes per Package
SDP
Catalog No. 50476571
Encompass
Unique design PCR tube strips feature an optically clear flat top for quantitative real time PCR techniques. These tubes are available in the standard 0.2 ml or with a new low profile design with a working volume of 40 ul. The flat top lid also provides a useful writing area when using tubes for their purposes. Supplied with tubes with attached cap per strip.
Catalog No. 50-476-571 Supplier Research Products International Corp Supplier No. 249941
Unique design PCR tube strips feature an optically clear flat top for quantitative real time PCR techniques. These tubes are available in the standard 0.2 ml or with a new low profile design with a working volume of 40 ul. The flat top lid also provides a useful writing area when using tubes for their purposes. Supplied with tubes with attached cap per strip.