Learn More
List Biological Laboratories Inc SNAPtide Peptide Substrate, Fluorogenic FRET Technology, Botulinum Neurotoxin Detection, 200 nmoles
SDP
Catalog No. NC2941191
Encompass_Preferred
The SNAPtide Peptide Substrate (o-Abz/Dnp) is a synthetic peptide aimed at detecting Clostridium botulinum Type A Neurotoxin activity using advanced FRET technology. This product offers a unique advantage in its design, allowing for precise enzymatic activity measurement through the release of fluorophores upon cleavage. Supplied as a lyophilized powder, it is well-suited for research applications and guarantees a purity of ≥95%.
Catalog No. NC2941191 Supplier List Biological Laboratories Inc Supplier No. 520
The SNAPtide Peptide Substrate (o-Abz/Dnp) is a synthetic peptide aimed at detecting Clostridium botulinum Type A Neurotoxin activity using advanced FRET technology. This product offers a unique advantage in its design, allowing for precise enzymatic activity measurement through the release of fluorophores upon cleavage. Supplied as a lyophilized powder, it is well-suited for research applications and guarantees a purity of ≥95%.