Learn More
Cayman Chemical (S)-N-Boc-L-Homoserine ethyl ester, Purity: ≥95%, CAS: 147325-09-5, 50 mg
SDP
Catalog No. 502991309
Encompass_Preferred
A synthetic intermediate in the synthesis of unsaturated caprolactams and monomer units for oxy-peptide nucleic acids from the starting material L-homoserine
Catalog No. 50-299-1309 Supplier Cayman Chemical Supplier No. 3054750 MG
A synthetic intermediate in the synthesis of unsaturated caprolactams and monomer units for oxy-peptide nucleic acids from the starting material L-homoserine