Learn More
Enzo Life Sciences SNC80 (25mg). CAS: 156727-74-1
SDP
Supplier:  Enzo Life Sciences ALX550172M025
Encompass_Preferred
Highly selective and potent non-peptide δ−opioid receptor agonist. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol, methanol, DMSO or dichloromethane. Long Term Storage: +4°C.
Catalog No. 50-200-9491