Learn More
AdipoGen SNC80 (5 mg)
SDP
Supplier:  AdipoGen AGCR10017M005
Encompass_Preferred
Chemical. CAS: 156727-74-1. Formula: C28H39N3O2. MW: 449.6. Highly selective and potent non-peptide delta-opioid receptor agonist. Mediates downregulation of the delta-opioid receptor. Shows antidepressant-like effects. Modulates voltage-dependent sodium channels. Antinociceptive. Activates ATP-sensitive K+ channels. Enhances amphetamine-mediated dopamine efflux.
Catalog No. 50-205-3763