Learn More
Abcam SNC80, delta opioid receptor agonist, 10MG
Catalog No. 502707355
Encompass_Preferred
MW 449.64 g/mol, Purity >99%. Highly potent and selective δ opioid receptor agonist (Ki = 9.4 nM). Centrally active following systemic administration. Displays antidepressant activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7355 Supplier Abcam Supplier No. AB12068410MG
MW 449.64 g/mol, Purity >99%. Highly potent and selective δ opioid receptor agonist (Ki = 9.4 nM). Centrally active following systemic administration. Displays antidepressant activity.
The product is subject to the following: Abcam Restricted Use Statement