Learn More
Aviva Systems Biology Corp SNCA Antibody - middle region (ARP42350_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42350_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNCA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SNCA. Alternate names: MGC110988, NACP, PARK1, PARK4, PD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58193