Learn More
Cayman Chemical Sancycline (hydrochloride), Purity: ≥95%, CAS: 6625-20-3, 100 mg
SDP
Catalog No. 502941769
Encompass_Preferred
A semisynthetic tetracycline antibiotic; more active than tetracycline against 339 strains of anaerobic bacteria (MIC90s = 1 and 32 μg/ml, respectively); active against tetracycline-resistant E. coli, S. aureus, and E. faecalis strains (MICs = 0.06-1 μg/ml); active against S. aureus in mice (ED50s = 0.46 and 0.6 mg/kg for intravenous and subcutaneous administration, respectively)
Catalog No. 50-294-1769 Supplier Cayman Chemical Supplier No. 28118100 MG
A semisynthetic tetracycline antibiotic; more active than tetracycline against 339 strains of anaerobic bacteria (MIC90s = 1 and 32 μg/ml, respectively); active against tetracycline-resistant E. coli, S. aureus, and E. faecalis strains (MICs = 0.06-1 μg/ml); active against S. aureus in mice (ED50s = 0.46 and 0.6 mg/kg for intravenous and subcutaneous administration, respectively)