Learn More
Medchemexpress LLC HY-136175 5mg Medchemexpress, SNDX-5613 CAS:2169919-21-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1361755MG
Encompass_Preferred
Medchemexpress, HY-136175 5mg SNDX-5613 CAS:2169919-21-3 SNDX-5613 is a potent and specific Menin-MLL inhibitor with a binding Ki of 0.149 nM and a cell based IC50 of 10-20 nM. SNDX-5613 can be used for the research of MLL-rearranged (MLL-r) acute leukemias, including acute lymphoblastic leukemia (ALL) and acute myeloid leukemia (AML). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9011