Learn More
Aviva Systems Biology Corp SNF1LK Antibody - N-terminal region (ARP45918_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45918_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SNF1LK. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SNF1LK. Alternate names: MSK, SIK, SNF1LKApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59166