Learn More
Metrohm USA 11 mL polypropylene vials, conical bottom, 1000 pack, no cap, 108 mm height, 16 mm outer diameter
SDP
Supplier:  Metrohm USA SNGIC3100V
Encompass
11 mL polypropylene vials, conical bottom, 1000 pack, no cap, 108 mm height, 16 mm outer diameter
Catalog No. NC3365660