Learn More
Cayman Chemical Sanggenone C, Purity: ≥98%, CAS: 80651-76-9, 10 mg
SDP
Catalog No. 502991158
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits TNF-α- or IL-1β-induced PMN adhesion to HSCs (IC50s = 27.29 and 54.43 nM, respectively); inhibits NF-κB activation in HSCs; induces apoptosis and production of ROS in HT-29 cells from 10-40 µM; decreases cell viability of HT-29 cells in vitro and reduces tumor growth in an HT-29 mouse xenograft model at 10 mg/kg; increases vertebrate column bone mineralization in a zebrafish model of prednisone-induced osteoporosis; attenuates cardiac hypertrophy and fibrosis and reduces activation of NFAT2 in a mouse model of pressure overload-induced cardiac hypertrophy
Catalog No. 50-299-1158 Supplier Cayman Chemical Supplier No. 3006110 MG
A flavonoid with diverse biological activities; inhibits TNF-α- or IL-1β-induced PMN adhesion to HSCs (IC50s = 27.29 and 54.43 nM, respectively); inhibits NF-κB activation in HSCs; induces apoptosis and production of ROS in HT-29 cells from 10-40 µM; decreases cell viability of HT-29 cells in vitro and reduces tumor growth in an HT-29 mouse xenograft model at 10 mg/kg; increases vertebrate column bone mineralization in a zebrafish model of prednisone-induced osteoporosis; attenuates cardiac hypertrophy and fibrosis and reduces activation of NFAT2 in a mouse model of pressure overload-induced cardiac hypertrophy