Learn More
Electron Microscopy Sciences ProPette LE™ Single Channel Pipette, 10-100 µl
SDP
Catalog No. 501931065
Encompass_Preferred
The ProPette LE is the next generation of high precision liquid handling instruments. It features improved actuation forces and a nearly 20% reduction in overall weight. It is an advanced pipette that exhibits superior accuracy and ergonomics and is comfortable to use, even during prolonged and repetitive pipetting.
The ProPette series features a continuously adjustable, digital micrometer for volume selection and a non-mandatory volume lock with simple one-handed activation/deactivation. The finely polished stainless piston and chemically resistant, maintenance-free seals provide unsurpassed accuracy. ProPette LE is fully autoclavable.
Range: 10-100 µl
Accuracy: ±7.5 - 1.0%
Reproducibility: ≤2.00- 0.30%
Catalog No. 50-193-1065 Supplier Electron Microscopy Sciences Supplier No. 7096105
The ProPette LE is the next generation of high precision liquid handling instruments. It features improved actuation forces and a nearly 20% reduction in overall weight. It is an advanced pipette that exhibits superior accuracy and ergonomics and is comfortable to use, even during prolonged and repetitive pipetting.
The ProPette series features a continuously adjustable, digital micrometer for volume selection and a non-mandatory volume lock with simple one-handed activation/deactivation. The finely polished stainless piston and chemically resistant, maintenance-free seals provide unsurpassed accuracy. ProPette LE is fully autoclavable.
Range: 10-100 µl
Accuracy: ±7.5 - 1.0%
Reproducibility: ≤2.00- 0.30%