Learn More
Electron Microscopy Sciences ProPette LE™ Single Channel Pipette, 1-5 ml
SDP
Supplier:  Electron Microscopy Sciences 7096109
Encompass_Preferred
The ProPette LE is the next generation of high precision liquid handling instruments. It features improved actuation forces and a nearly 20% reduction in overall weight. It is an advanced pipette that exhibits superior accuracy and ergonomics and is comfortable to use, even during prolonged and repetitive pipetting.
The ProPette series features a continuously adjustable, digital micrometer for volume selection and a non-mandatory volume lock with simple one-handed activation/deactivation. The finely polished stainless piston and chemically resistant, maintenance-free seals provide unsurpassed accuracy. ProPette LE is fully autoclavable.
Range: 1-5 ml
Accuracy: ±2.5 - 0.8%
Reproducibility: ≤1.00- 0.15%
Catalog No. 50-193-1069