Learn More
Integra Biosciences Corp Single Channel EVOLVE Pipettes, 2 – 20 ul
SDP
Catalog No. NC1331649
Encompass
Adjust volumes more than ten times faster thanks to three adjustable dials. Two differently sized plunger springs allow users to customize the force needed to depress the plunger to their preference. GripTips are always firmly attached, perfectly aligned and feature a low attachment and ejection force.
This item is not returnable. View return policy
Catalog No. NC1331649 Supplier Integra Biosciences Corp Supplier No. 3013
Adjust volumes more than ten times faster thanks to three adjustable dials. Two differently sized plunger springs allow users to customize the force needed to depress the plunger to their preference. GripTips are always firmly attached, perfectly aligned and feature a low attachment and ejection force.