Learn More
MSC Miller AirCore Full-Body Harness, Aluminum Hardware, Quick-Connect Buckles, Universal, 1 ea
Catalog No. 5000507557
Encompass
The AirCore full-body harness is a premium fall protection solution engineered for demanding high-risk tasks. This harness utilizes open-core padding technology that is breathable to maximize airflow and minimize heat and moisture buildup. Constructed with stretchable polyester webbing and lightweight aluminum hardware, it ensures superior flexibility, mobility, and corrosion resistance. The contoured ergonomic design reduces contact points around the neck and thighs to prevent chafing, while the quick-connect chest and leg buckles facilitate easy donning.
  • Features breathable, open-core padding for comfort
  • Includes patented stretchable webbing for increased mobility
  • Equipped with lightweight, corrosion-resistant aluminum hardware
  • Supports a weight capacity of 400 lbs.
  • Designed with a contoured shape to minimize irritation
  • Meets ANSI, CSA, and OSHA safety standards
Catalog No. 50-005-07557 Supplier MSC Supplier No. 80891351
The AirCore full-body harness is a premium fall protection solution engineered for demanding high-risk tasks. This harness utilizes open-core padding technology that is breathable to maximize airflow and minimize heat and moisture buildup. Constructed with stretchable polyester webbing and lightweight aluminum hardware, it ensures superior flexibility, mobility, and corrosion resistance. The contoured ergonomic design reduces contact points around the neck and thighs to prevent chafing, while the quick-connect chest and leg buckles facilitate easy donning.
  • Features breathable, open-core padding for comfort
  • Includes patented stretchable webbing for increased mobility
  • Equipped with lightweight, corrosion-resistant aluminum hardware
  • Supports a weight capacity of 400 lbs.
  • Designed with a contoured shape to minimize irritation
  • Meets ANSI, CSA, and OSHA safety standards