Learn More
MSC Miller By Honeywell Aircore Full Body Harness, Vest-Style, Stretchable, Small/Medium, 400 lb, 1 Each
Catalog No. 5000507688
Encompass
This full body harness is designed for general industry fall protection applications. It features lightweight, stretchable construction to enhance comfort and mobility while providing a high weight capacity for demanding work environments. The harness is engineered with breathable padding technology for moisture control and includes integrated features for fall indication.
  • Features stretchable webbing for increased movement
  • Includes breathable open-core padding for comfort
  • Equipped with quick-connect chest buckles and tongue leg strap buckles
  • Utilizes durable aluminum hardware
  • Includes fall indicator on the back pad
  • Designed with a subpelvic strap
  • Meets ANSI A10.32, ANSI Z359.1, and OSHA 1926.502 standards
  • Provides a 400 lb load capacity
Catalog No. 50-005-07688 Supplier MSC Supplier No. 80891310
This full body harness is designed for general industry fall protection applications. It features lightweight, stretchable construction to enhance comfort and mobility while providing a high weight capacity for demanding work environments. The harness is engineered with breathable padding technology for moisture control and includes integrated features for fall indication.
  • Features stretchable webbing for increased movement
  • Includes breathable open-core padding for comfort
  • Equipped with quick-connect chest buckles and tongue leg strap buckles
  • Utilizes durable aluminum hardware
  • Includes fall indicator on the back pad
  • Designed with a subpelvic strap
  • Meets ANSI A10.32, ANSI Z359.1, and OSHA 1926.502 standards
  • Provides a 400 lb load capacity