Learn More
MSC Miller AirCore Vest-Style Harness, Tongue Buckle, Small/Medium, 1 ea
Catalog No. 5000507681
Encompass
This vest-style harness is designed for reliable fall protection in demanding environments. Featuring a stretchable design and integrated back padding, it offers enhanced comfort and mobility for the wearer during extended use. The harness is equipped with five points of adjustment to ensure a secure and customizable fit.
  • Features a durable construction with polyester webbing
  • Includes back and side D-ring locations for versatile attachment
  • Equipped with tongue-style leg strap buckles
  • Utilizes quick-connect chest strap buckles
  • Supports a capacity of 400 lb
  • Meets ANSI A10.32, ANSI Z359.1, and OSHA 1926.502 standards
Catalog No. 50-005-07681 Supplier MSC Supplier No. 80890783
This vest-style harness is designed for reliable fall protection in demanding environments. Featuring a stretchable design and integrated back padding, it offers enhanced comfort and mobility for the wearer during extended use. The harness is equipped with five points of adjustment to ensure a secure and customizable fit.
  • Features a durable construction with polyester webbing
  • Includes back and side D-ring locations for versatile attachment
  • Equipped with tongue-style leg strap buckles
  • Utilizes quick-connect chest strap buckles
  • Supports a capacity of 400 lb
  • Meets ANSI A10.32, ANSI Z359.1, and OSHA 1926.502 standards