Learn More
Innovative Research Inc Sngl Dnr Hmn Nasal Fluid Vial
Catalog No. 502744037
Encompass_Preferred
Single Donor Human Nasal Fluid Vial - Post COVID - Single Donor Human Nasal Fluid from Innovative Research is collected from a unique single donor and frozen immediately after collection. One vial is greater than or equal to approximately 0.5ml. Human nasal fluid is rich in glycoproteins and water, and is typically produced from cells found in mucous glands.
Catalog No. 50-274-4037 Supplier Innovative Research Inc Supplier No. IRHUNFSVIAL
Add to Cart
Add to Cart
Single Donor Human Nasal Fluid Vial - Post COVID - Single Donor Human Nasal Fluid from Innovative Research is collected from a unique single donor and frozen immediately after collection. One vial is greater than or equal to approximately 0.5ml. Human nasal fluid is rich in glycoproteins and water, and is typically produced from cells found in mucous glands.