Learn More
Innovative Research Inc Sngl Dnr Hmn Obesity Pla
Catalog No. 502744097
Encompass_Preferred
Single Donor Human Obesity Plasma - Single Donor Human Obesity Plasma from Innovative Research is whole blood derived. Available volumes are variable, and range from 0.5ml to 1.0ml per vial (you can select your exact samples from our inventory, if desired). A detailed patient history and clinical information is recorded at the time of draw, and multiple samples and plasma/serum matched sets from the same donor are often available.
Catalog No. 50-274-4097 Supplier Innovative Research Inc Supplier No. IPLASOBS
Single Donor Human Obesity Plasma - Single Donor Human Obesity Plasma from Innovative Research is whole blood derived. Available volumes are variable, and range from 0.5ml to 1.0ml per vial (you can select your exact samples from our inventory, if desired). A detailed patient history and clinical information is recorded at the time of draw, and multiple samples and plasma/serum matched sets from the same donor are often available.