Learn More
Innovative Research Inc Sngl Dnr Hmn Seminal Fluid 1ml
Catalog No. 502744029
Encompass_Preferred
Single Donor Human Seminal Fluid 1ml - Single Donor Human Seminal Fluid from Innovative Research is whole semen collected and centrifuged to separate the fluid from the sperm cells. There are no preservatives in this product.
Catalog No. 50-274-4029 Supplier Innovative Research Inc Supplier No. IRHUSSMF1ML
Single Donor Human Seminal Fluid 1ml - Single Donor Human Seminal Fluid from Innovative Research is whole semen collected and centrifuged to separate the fluid from the sperm cells. There are no preservatives in this product.