Learn More
Innovative Research Inc Sngl Dnr Hmn Slva 5ml
Catalog No. 502744033
Encompass_Preferred
Single Donor Human Saliva 5ml - Post COVID - Single Donor Human Saliva from Innovative Research is collected from a single unique donor. Produced in the salivary glands, human saliva is 99.5% water, but contains many important substances including electrolytes, mucus, antibacterial compounds, and various enzymes. Human Saliva donors by gender and race are available upon request. This product is collected per-order, and can be sent to you fresh or frozen. If you have a preference, please mention it in the Specifications box above. This product will ship frozen unless otherwise indicated.
Catalog No. 50-274-4033 Supplier Innovative Research Inc Supplier No. IRHUSLS5ML
Single Donor Human Saliva 5ml - Post COVID - Single Donor Human Saliva from Innovative Research is collected from a single unique donor. Produced in the salivary glands, human saliva is 99.5% water, but contains many important substances including electrolytes, mucus, antibacterial compounds, and various enzymes. Human Saliva donors by gender and race are available upon request. This product is collected per-order, and can be sent to you fresh or frozen. If you have a preference, please mention it in the Specifications box above. This product will ship frozen unless otherwise indicated.