Learn More
Innovative Research Inc Sngl Dnr Hmn Slva Age 3-9 1
Catalog No. 502744030
Encompass_Preferred
Single Donor Human Pediatric Saliva Ages 3-9 1 Sample - Single Donor Human Pediatric Saliva from Innovative Research is a sample (approximately 1 mL) of pediatric saliva. This may be purchased within age groups 3-9 years of age and 10-17 years of age. Parental approval received prior to collection.
Catalog No. 50-274-4030 Supplier Innovative Research Inc Supplier No. IRHUSSLPED391
Single Donor Human Pediatric Saliva Ages 3-9 1 Sample - Single Donor Human Pediatric Saliva from Innovative Research is a sample (approximately 1 mL) of pediatric saliva. This may be purchased within age groups 3-9 years of age and 10-17 years of age. Parental approval received prior to collection.