Learn More
Innovative Research Inc Sngl Dnr Hmn Urn 100ml
Supplier:  Innovative Research Inc IRHUURES100ML
Encompass_Preferred
Single Donor Human Urine 100ml - Single Donor Human Urine from Innovative Research is a liquid product of the body that is secreted by the kidneys and excreted through the urethra. Cellular metabolism generates numerous waste compounds, many rich in nitrogen, which require elimination from the bloodstream. This waste is eventually expelled from the body in a process known as micturition, the primary method for excreting water-soluble chemicals from the body. These chemicals can be detected and analyzed by urinalysis. Aside from water, urine contains an assortment of inorganic salts and organic compounds, including proteins, hormones, and a wide range of metabolites. Single Donor Human Urine can be obtained by age, gender, and race if requested. This product is available in Fresh or Frozen forms. If you have a preference, please indicate in the specifications field above. This product will ship frozen unless otherwise indicated.
Catalog No. 50-274-4019