Learn More
Cpi International SNGL ELEMENT ARSENIC STD
SDP
Catalog No. NC9177663
Encompass
Single-Element Standards 1000microgram/mL, Element: Arsenic, Source: As metal, Purity: 99.999 + %, Matrix: 2% HNO3
Catalog No. NC9177663 Supplier Cpi International Supplier No. S4400100031
Add to Cart
Add to Cart
Single-Element Standards 1000microgram/mL, Element: Arsenic, Source: As metal, Purity: 99.999 + %, Matrix: 2% HNO3