Learn More
Med Vet International Single Frosted Slides, Enhanced
Catalog No. 503045138
Encompass_Preferred
Premiere premium grade slides provide consistent results for all laboratory tests. The edges are also finely polished to produce high-quality smears and clipped corners make them easy and safe to handle. Premiere name printed on color slides indicates frosted side. Packed 72pcs/box, 20 boxes (10gross) per case. Order per gross in multiples of 10 gross.
Catalog No. 50-304-5138 Supplier Med Vet International Supplier No. 9105E
Premiere premium grade slides provide consistent results for all laboratory tests. The edges are also finely polished to produce high-quality smears and clipped corners make them easy and safe to handle. Premiere name printed on color slides indicates frosted side. Packed 72pcs/box, 20 boxes (10gross) per case. Order per gross in multiples of 10 gross.