Learn More
Innovative Research Inc Sngl Hmn Covid Pos Pla 100ul
Catalog No. 502744108
Encompass_Preferred
Single Donor Human SARS-Cov-2 Positive Plasma 100ul - Single Donor Human SARS-Cov-2 Positive Plasma from Innovative Research is a whole blood derived product processed using EDTA as the anticoagulant. Samples were collected through an IRB-approved protocol with full donor consent, and COVID-19 positive status has been confirmed via SARS-CoV-2 IgG antibody assay and/or PCR positive test results. Please note that this product will contain differing levels of IgM, IgA, and/or IgG antibody levels from lot to lot as this is a single donor product. This plasma is suitable for use as a positive control and to confirm antibody assay results, among other research applications.
Catalog No. 50-274-4108 Supplier Innovative Research Inc Supplier No. IPLASCOV2P100UL
Single Donor Human SARS-Cov-2 Positive Plasma 100ul - Single Donor Human SARS-Cov-2 Positive Plasma from Innovative Research is a whole blood derived product processed using EDTA as the anticoagulant. Samples were collected through an IRB-approved protocol with full donor consent, and COVID-19 positive status has been confirmed via SARS-CoV-2 IgG antibody assay and/or PCR positive test results. Please note that this product will contain differing levels of IgM, IgA, and/or IgG antibody levels from lot to lot as this is a single donor product. This plasma is suitable for use as a positive control and to confirm antibody assay results, among other research applications.