Learn More
Innovative Research Inc Sngl Hmn Pla Bld Der Li Hep
Catalog No. 502744104
Encompass_Preferred
Single Donor Human Plasma (Blood Derived) Li Heparin 50ml - Single Donor Human Plasma from Innovative Research is whole blood derived. The whole blood is collected from donors in an FDA-approved collection center and processed into plasma via centrifugation. The plasma is frozen immediately after processing. Each unit is tested for the standard FDA-required viral markers, and found negative using FDA-approved methods. Innovative Research has a wide network of FDA-approved collection centers across the United States, which allows us to collect from nearly any donor type. We are able to collect this product using the anticoagulant that will work best for your research. Our standard anticoagulant options include K2 EDTA, K3 EDTA, Na EDTA, Na Citrate, Na Heparin, and Li Heparin, with others available upon request.
Catalog No. 50-274-4104 Supplier Innovative Research Inc Supplier No. IPLASLIH50ML
Single Donor Human Plasma (Blood Derived) Li Heparin 50ml - Single Donor Human Plasma from Innovative Research is whole blood derived. The whole blood is collected from donors in an FDA-approved collection center and processed into plasma via centrifugation. The plasma is frozen immediately after processing. Each unit is tested for the standard FDA-required viral markers, and found negative using FDA-approved methods. Innovative Research has a wide network of FDA-approved collection centers across the United States, which allows us to collect from nearly any donor type. We are able to collect this product using the anticoagulant that will work best for your research. Our standard anticoagulant options include K2 EDTA, K3 EDTA, Na EDTA, Na Citrate, Na Heparin, and Li Heparin, with others available upon request.