Learn More
MSC King Electric Line Voltage Wall Thermostat, 22 A, 120-240 V, 41°F to 90°F, 1 ea
Catalog No. 5000509133
Encompass
This line voltage wall thermostat provides reliable temperature control for heating systems. Designed for ease of use, it features a bi-metal mechanical sensing element and allows for precise manual adjustments via a large control knob. The unit is engineered for installation flexibility and operates within a standard range, making it suitable for various residential and commercial heating applications.
  • Temperature range of 41°F to 90°F
  • Bi-metal mechanical sensing element with reduced temperature swing
  • Compatible with 120V, 208V, 240V, and 277V systems
  • Rated for 25 A
  • Features control dials in °C and °F
  • Mounts directly to a 2x4-in. vertical electrical outlet box
  • No ground wire design for simplified installation
  • ETL certified
Catalog No. 50-005-09133 Supplier MSC Supplier No. 96894589
This line voltage wall thermostat provides reliable temperature control for heating systems. Designed for ease of use, it features a bi-metal mechanical sensing element and allows for precise manual adjustments via a large control knob. The unit is engineered for installation flexibility and operates within a standard range, making it suitable for various residential and commercial heating applications.
  • Temperature range of 41°F to 90°F
  • Bi-metal mechanical sensing element with reduced temperature swing
  • Compatible with 120V, 208V, 240V, and 277V systems
  • Rated for 25 A
  • Features control dials in °C and °F
  • Mounts directly to a 2x4-in. vertical electrical outlet box
  • No ground wire design for simplified installation
  • ETL certified