Learn More
REPLIGEN CORPORATION Repligen ProConnex Single Use Clamp, Ergonomic Design, Overtightening Prevention, 10 PK
SDP
Catalog No. NC1659673
Encompass
The Repligen ProConnex Single Use Hygienic Plastic Clamp is designed to provide secure connections in bioprocess assemblies. This clamp features an ergonomic design for easy single-handed operation, preventing overtightening and reducing failure risks. Compliant with bioprocess quality and ASME-BPE standards, it ensures reliability in both metallic and non-metallic systems, making it an essential tool for efficient modular assembly operations.
Catalog No. NC1659673 Supplier REPLIGEN CORPORATION Supplier No. BFS1500010
The Repligen ProConnex Single Use Hygienic Plastic Clamp is designed to provide secure connections in bioprocess assemblies. This clamp features an ergonomic design for easy single-handed operation, preventing overtightening and reducing failure risks. Compliant with bioprocess quality and ASME-BPE standards, it ensures reliability in both metallic and non-metallic systems, making it an essential tool for efficient modular assembly operations.