Learn More
REPLIGEN CORPORATION Clip Clamp For 1/2″ & 3/4″ Hygienic Clamp. Tubing Management System
SDP
Catalog No. NC1659674
Encompass
The BioFlex Clip Clamp allows system designers to create a complete Tubing Management System (TMS) for single use tubing systems when combined with the BFS-075 TC Clamp. It is the first complete system that helps manage, identify and organize single use fluid paths. Patent Pending.
Catalog No. NC1659674 Supplier REPLIGEN CORPORATION Supplier No. BFS075010
The BioFlex Clip Clamp allows system designers to create a complete Tubing Management System (TMS) for single use tubing systems when combined with the BFS-075 TC Clamp. It is the first complete system that helps manage, identify and organize single use fluid paths. Patent Pending.